Triethyltin chloride

Danger

Synonyms: Chlorotriethylstannane

39601-14 39601-14
AA39601-14EA 391.69 USD
CAAA39601-14 AA39601-14
Triethyltin chloride
Triethyltin chloride
Formula: C₆H₁₅ClSn
MW: 241.35 g/mol
Boiling Pt: 206 °C
Density: 1,429
Storage Temperature: Ambient
MDL Number: MFCD00040414
CAS Number: 994-31-0
EINECS: 213-616-1
UN: 2788
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR