Barium fluoride, crystal optic rectangle 30 mm x 15 mm x 4 mm, polished both sides
Supplier: Thermo Scientific Chemicals
|
Warning
|
038459-KT
38459-KT
AA38459-KTEA
273.99
USD
AA38459-KT
CAAA38459-KT
Barium fluoride, crystal optic rectangle 30 mm x 15 mm x 4 mm, polished both sides
Barium fluoride
Common Applications: Spectroscopy, IR window
Formula:
BaF₂ MW: 175.33 g/mol Boiling Pt: 2260 °C (1013 hPa) Melting Pt: 1270 °C Density: 4.89 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003450 CAS Number: 7787-32-8 EINECS: 232-108-0 UN: 1564 ADR: 6.1,III Merck Index: 12,01003 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...