Ethanol-[D₆] (99.5% D) (Isotopic) for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ethyl alcohol-D6 Hydroxy ethane-D6 Methyl carbinol-D6 Ethanol-[²H₆]

36502-06 036502-06
AA36502-06EA 437.99 USD
AA36502-06 CAAA36502-06
Ethanol-[D₆] (99.5% D) (Isotopic) for NMR spectroscopy
Ethanol-[D6]
Formula: D₃CCD₂OD
MW: 52.02 g/mol
Boiling Pt: 79 °C
Melting Pt: –114.5 °C
Density: 0.91
Flash Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00051020
CAS Number: 1516-08-1
EINECS: 216-162-2
UN: 1170
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR