Chromium(VI) oxide 98+% ACS

Danger

Synonyms: Chromic anhydride Chromium trioxide

36468-36 36468-22
AA36468-22EA 66.2 USD
AA36468-22 AA36468-36
Chromium(VI) oxide 98+% ACS
Chromium (VI) oxide

Chromium plating, copper stripping, aluminum anodizing, corrosion inhibitor, photography, ceramic glazes, colored glass, oxidizing agent in organic chemistry

Formula: CrO₃
MW: 99.99 g/mol
Boiling Pt: 250 °C (dec.)
Melting Pt: 196 °C
Density: 2.7
Storage Temperature: Ambient
MDL Number: MFCD00010952
CAS Number: 1333-82-0
EINECS: 215-607-8
UN: 1463
ADR: 5.1,II
REACH: 01-2119458868-17
Merck Index: 13,02256

Order Now

Specification Test Results

Insoluble matter 0.01%
Cl 0.005%
NO6 0.03%
SO3 0.005%
Na 0.2%
Al 0.02%
Ba 0.01%
Fe 0.02%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR