Mercury(I) chloride ≥99.5% ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dimercury dichloride Mercurous chloride

36419-22 036419-22
AA36419-22EA 92.5 USD
CAAA36419-22 AA36419-22
Mercury(I) chloride ≥99.5% ACS
Mercury (I) chloride
Formula: Cl₂Hg₂
MW: 472.09 g/mol
Boiling Pt: 384 °C (1013 hPa)
Melting Pt: 400 °C (subl.)
Density: 7.15
Storage Temperature: Ambient
MDL Number: MFCD00011043
CAS Number: 10112-91-1
EINECS: 233-307-5
UN: 2025
ADR: 6.1,III
Merck Index: 12,05951

Order Now

Specification Test Results

Residue after reduction 0.02%
HgCl2 0.01%
SO3 0.01%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR