Sodium hexafluoroaluminate ≥99.5% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Trisodium hexafluoroaluminate
36345-18
036345-30
036345-18
36345-30
AA36345-30EA
411.99
USD
AA36345-30
CAAA36345-30
AA36345-18
CAAA36345-18
Sodium hexafluoroaluminate ≥99.5% (metals basis)
Sodium hexafluoroaluminate
Formula:
AlF₆Na₃ MW: 209.94 g/mol Melting Pt: 1000 °C Density: 2.90 Storage Temperature: Ambient |
MDL Number:
MFCD00003507 CAS Number: 13775-53-6 EINECS: 237-410-6 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...