Zinc diarsenide ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals

Danger

Synonyms: Zinc diarsenide

36189-09 36189-09
AA36189-09EA 211 USD
AA36189-09 CAAA36189-09
Zinc diarsenide ≥99.999% (metals basis)
Zinc diarsenide
Formula: As₂Zn
MW: 215.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00151429
CAS Number: 12044-55-2
EINECS: 234-956-7
UN: 1557
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR