Calcium metasilicate ≥99% (metals basis)

Warning

Synonyms: Calcium silicate

35843-36 35843-14
AA35843-36EA 757.02 USD
AA35843-36 AA35843-14
Calcium metasilicate ≥99% (metals basis)
Calcium metasilicate
Formula: CaO₃Si
MW: 116.16 g/mol
Melting Pt: 1540 °C
Density: 2,905
Storage Temperature: Ambient
MDL Number: MFCD00015979
CAS Number: 10101-39-0
EINECS: 233-250-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR