Phosphorus(V) fluoride

Danger

33543-22 33543-22
AA33543-22EA 1255.41 USD
CAAA33543-22 AA33543-22
Phosphorus(V) fluoride
Phosphorus (V) fluoride
Formula: F₅P
MW: 125.97 g/mol
Boiling Pt: –84.6 °C
Melting Pt: –93.8 °C
Density: 5,805
Storage Temperature: Ambient
MDL Number: MFCD00040326
CAS Number: 7647-19-0
EINECS: 231-602-3
UN: 2198
ADR: 2.3,
Merck Index: 12,07509

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR