2-Methylpyrrolidine 97%

Supplier: Thermo Scientific Chemicals
Danger

31902-03 31902-06 031902-03 031902-06
AA31902-03EA 81.49 USD
AA31902-03 CAAA31902-03 AA31902-06 CAAA31902-06
2-Methylpyrrolidine 97%
2-Methylpyrrolidine
Formula: C₅H₁₁N
MW: 85.15 g/mol
Density: 0.834
Flash Pt: 2 °C (37 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014491
CAS Number: 765-38-8
EINECS: 212-144-3
UN: 2733
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR