Boron trifluoride 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Boron fluoride

22874-22 022874-22
AA22874-22EA 1227.99 USD
CAAA22874-22 AA22874-22
Boron trifluoride 99+%
Boron trifluoride
Formula: BF₃
MW: 67.81 g/mol
Boiling Pt: –100 °C
Melting Pt: –127 °C
Density: 3,076
Storage Temperature: Ambient
MDL Number: MFCD00011316
CAS Number: 7637-07-2
EINECS: 237-383-0
UN: 1008
ADR: 2.3,
Merck Index: 12,01379

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR