Zirconium tetraiodide 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Zirconium (IV) iodide

17400-14 017400-14
AA17400-14EA 650 USD
CAAA17400-14 AA17400-14
Zirconium tetraiodide 99%
Zirconium tetraiodide
Formula: I₄Zr
MW: 598.84 g/mol
Melting Pt: 431 °C (subl.)
Density: 4,85
Storage Temperature: Ambient
MDL Number: MFCD00049641
CAS Number: 13986-26-0
EINECS: 237-780-9
UN: 3260
ADR: 8,III
Merck Index: 14,10178

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR