1-Chloroadamantane 98%

Supplier: Thermo Scientific Chemicals

15169-06 15169-14 15169-06 15169-14 15169-22
AA15169-22EA 327.75 USD
CAAA15169-14 AA15169-22 AA15169-06 AA15169-14 CAAA15169-06
1-Chloroadamantane 98%
1-Chloroadamantane
Formula: C₁₀H₁₅Cl
Melting Pt: 165 °C
Density: 1,26 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00075627
CAS Number: 935-56-8
EINECS: 213-305-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR