Iron 99+% (metals basis), wire, Diameter 2.0 mm (0.08 in)
Supplier: Thermo Scientific Chemicals
14246-H2
014246-H2
AA14246-H2EA
666.99
USD
AA14246-H2
CAAA14246-H2
Iron 99+% (metals basis), wire, Diameter 2.0 mm (0.08 in)
Iron
Formula:
Fe MW: 55.85 g/mol Boiling Pt: 3000 °C (1013 hPa) Melting Pt: 1535 °C Density: 7.86 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010999 CAS Number: 7439-89-6 EINECS: 231-096-4 Merck Index: 13,05109 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...