Aluminium ≥99% (metals basis), foil [Si & Fe (combined) ∼ 1%], annealed, Thickness 1.0 mm (0.04 in)

Supplier: Thermo Scientific Chemicals

Synonyms: Aluminum

14192-RV 14192-VV 014192-RV 14192-VK 14192-VV
AA14192-RVEA 154.99 USD
AA14192-RV CAAA14192-VK CAAA14192-RV AA14192-VV CAAA14192-VV
Aluminium ≥99% (metals basis), foil [Si & Fe (combined) ∼ 1%], annealed, Thickness 1.0 mm (0.04 in)
Aluminium
Formula: Al
MW: 26.98 g/mol
Boiling Pt: 2327 °C (1013 hPa)
Melting Pt: 660 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR