Zirconium ≥99.2% (metals basis), sponge ≤ 25.4 mm (1.0 in)

Danger

14191-36 14191-22
AA14191-36EA 431.57 USD
AA14191-36 AA14191-22
Zirconium ≥99.2% (metals basis), sponge ≤ 25.4 mm (1.0 in)
Zirconium
Formula: Zr
MW: 91.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9
UN: 2008
ADR: 4.2,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR