Disodium fluorophosphate
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Sodium fluorophosphate
14129-36
014129-36
AA14129-36EA
626.99
USD
CAAA14129-36
AA14129-36
Disodium fluorophosphate
Disodium fluorophosphate
Formula:
FNa₂O₃P MW: 143.95 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00014248 CAS Number: 10163-15-2 EINECS: 233-433-0 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...