Cerium(IV) oxide 20% in water, colloidal dispersion

Danger

Synonyms: Ceric oxideCerium dioxide

12730-A3 012730-A3 12730-36 012730-36
AA12730-A3EA 351.99 USD
AA12730-A3 AA12730-36 CAAA12730-A3 CAAA12730-36
Cerium(IV) oxide 20% in water, colloidal dispersion
Cerium (IV) oxide
Formula: CeO₂
MW: 172.11 g/mol
Density: 1,22
Storage Temperature: Ambient
MDL Number: MFCD00010933
CAS Number: 1306-38-3
EINECS: 215-150-4
UN: 3264
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR