Thallium ≥99.999% (metals basis), rods, Diameter 12.7 mm (0.5 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
12714-0Y
12714-DO
AA12714-0YEA
2960.99
USD
AA12714-0Y
AA12714-DO
Thallium ≥99.999% (metals basis), rods, Diameter 12.7 mm (0.5 in)
Thallium
Formula:
Tl MW: 204.38 g/mol Boiling Pt: 1457 °C (1013 hPa) Melting Pt: 301 °C Density: 11.85 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134063 CAS Number: 7440-28-0 EINECS: 231-138-1 UN: 1707 ADR: 6.1,II Merck Index: 12,09391 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...