Potassium hexacyanoplatinate(IV)

Danger

Synonyms: Dipotassium hexacyanoplatinate

12678-MD 12678-03 12678-03
AA12678-MDEA 160.95 USD
AA12678-MD CAAA12678-03 AA12678-03
Potassium hexacyanoplatinate(IV)
Potassium hexacyanoplatinate (IV)
Formula: C₆K₂N₆Pt
MW: 429.38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00058745
CAS Number: 16920-94-8
EINECS: 240-978-8
UN: 1588
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR