Cesium perchlorate, REacton®, Reagent Grade

Supplier: Thermo Scientific Chemicals
Danger

12308-14 012308-14
AA12308-14EA 76.09 USD
CAAA12308-14 AA12308-14
Cesium perchlorate, REacton®, Reagent Grade
Cesium perchlorate
Formula: ClCsO₄
MW: 232.36 g/mol
Melting Pt: 250 °C (dec.)
Density: 3.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010956
CAS Number: 13454-84-7
EINECS: 236-643-0
UN: 1481
ADR: 5.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR