Dipotassium hexachloroiridate Ir ≥ 39%
|
Warning
|
Synonyms:
Potassium hexachloroiridate (IV)
011888-14
011888-03
11888-03
11888-06
11888-14
011888-06
AA11888-06EA
1493.99
USD
AA11888-06
CAAA11888-06
AA11888-14
AA11888-03
CAAA11888-03
CAAA11888-14
Dipotassium hexachloroiridate Ir ≥ 39%
Dipotassium hexachloroiridate
Formula:
Cl₆IrK₂ Melting Pt: Dec. Storage Temperature: Ambient |
MDL Number:
MFCD00011369 CAS Number: 16920-56-2 EINECS: 240-976-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...