Phosphoryl trichloride ≥99.999% (metals basis)
|
Danger
|
Synonyms:
Phosphorus (V) oxide chloride
11847-14
AA11847-14EA
150.99
USD
AA11847-14
Phosphoryl trichloride ≥99.999% (metals basis)
Phosphoryl trichloride
Formula:
POCl₃ MW: 153.33 g/mol Boiling Pt: 105.8 °C Melting Pt: 1.25 °C Density: 1.645 Storage Temperature: Ambient |
MDL Number:
MFCD00011443 CAS Number: 10025-87-3 EINECS: 233-046-7 UN: 1810 ADR: 6.1,I Merck Index: 12,07506 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...