Antimony(III) bromide ≥99.995% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Antimony tribromide
11686-09
011686-09
AA11686-09EA
123.99
USD
CAAA11686-09
AA11686-09
Antimony(III) bromide ≥99.995% (metals basis)
Antimony (III) bromide
Formula:
Br₃Sb MW: 361.47 g/mol Boiling Pt: 280 °C Melting Pt: 96.6 °C Density: 4.148 Storage Temperature: Ambient |
MDL Number:
MFCD00016317 CAS Number: 7789-61-9 EINECS: 232-179-8 UN: 3260 ADR: 8,II Merck Index: 12,00747 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...