Rhodium ≥99.8% (metals basis), wire, Diameter 0.127 mm (0.005 in)
11578-BU
011578-BY
011578-BU
11578-BY
AA11578-BUEA
432.99
USD
AA11578-BU
CAAA11578-BU
AA11578-BY
CAAA11578-BY
Rhodium ≥99.8% (metals basis), wire, Diameter 0.127 mm (0.005 in)
Rhodium
Formula:
Rh MW: 102.9 g/mol Boiling Pt: 3400…3700 °C (1013 hPa) Melting Pt: 1960 °C Density: 12,41 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011201 CAS Number: 7440-16-6 EINECS: 231-125-0 Merck Index: 12,08353 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...