Rhodium ≥99.8% (metals basis), wire, Diameter 0.25 mm (0.01 in)
11577-BY
011577-BU
011577-BY
11577-BU
AA11577-BUEA
336.99
USD
AA11577-BU
CAAA11577-BU
AA11577-BY
CAAA11577-BY
Rhodium ≥99.8% (metals basis), wire, Diameter 0.25 mm (0.01 in)
Rhodium
Formula:
Rh MW: 102.9 g/mol Boiling Pt: 3400…3700 °C (1013 hPa) Melting Pt: 1960 °C Density: 12,41 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011201 CAS Number: 7440-16-6 EINECS: 231-125-0 Merck Index: 12,08353 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...