Rhodium ≥99.8% (metals basis), wire, Diameter 1.0 mm (0.04 in)
11575-BP
011575-BS
011575-BP
11575-BS
AA11575-BPEA
500.99
USD
CAAA11575-BP
CAAA11575-BS
AA11575-BP
AA11575-BS
Rhodium ≥99.8% (metals basis), wire, Diameter 1.0 mm (0.04 in)
Rhodium
Formula:
Rh MW: 102.9 g/mol Boiling Pt: 3400…3700 °C (1013 hPa) Melting Pt: 1960 °C Density: 12,41 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011201 CAS Number: 7440-16-6 EINECS: 231-125-0 Merck Index: 12,08353 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...