Potassium hexafluoroarsenate ≥99% (metals basis)
|
Danger
|
Synonyms:
Potassium hexafluoroarsenate(V)
11532-06
11532-14
11532-14
11532-06
AA11532-06EA
51.84
USD
AA11532-06
CAAA11532-06
AA11532-14
CAAA11532-14
Potassium hexafluoroarsenate ≥99% (metals basis)
Potassium hexafluoroarsenate
Formula:
AsF₆K MW: 228.01 g/mol Melting Pt: 400 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042546 CAS Number: 17029-22-0 EINECS: 241-102-7 UN: 1557 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...