Cobalt ≥99.995% (metals basis), foil, Puratronic®, Thickness 2.0 mm (0.08 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
11449-FF
011449-FI
011449-FF
11449-FI
AA11449-FFEA
381.99
USD
AA11449-FF
CAAA11449-FF
AA11449-FI
CAAA11449-FI
Cobalt ≥99.995% (metals basis), foil, Puratronic®, Thickness 2.0 mm (0.08 in)
Cobalt
Formula:
Co MW: 58.93 g/mol Boiling Pt: 2870 °C (1013 hPa) Melting Pt: 1495 °C Density: 8.9 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010935 CAS Number: 7440-48-4 EINECS: 231-158-0 Merck Index: 12,02488 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...