Silver hard ≥99.9% (metals basis), foil, hard, Thickness 1.0 mm (0.04 in)

Supplier: Thermo Scientific Chemicals

011438-FI 11438-FF 11438-FI 011438-FF 011438-GH 11438-GH
AA11438-GHEA 652.99 USD
CAAA11438-FI AA11438-GH AA11438-FF AA11438-FI CAAA11438-GH CAAA11438-FF
Silver hard ≥99.9% (metals basis), foil, hard, Thickness 1.0 mm (0.04 in)
Silver
Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212 °C (1013 hPa)
Melting Pt: 962 °C
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR