Iridium ≥99.8% (metals basis), wire, Diameter 0.5 mm (0.02 in)
11430-BU
11430-BQ
011430-BQ
011430-BU
AA11430-BQEA
220.99
USD
CAAA11430-BQ
AA11430-BQ
CAAA11430-BU
AA11430-BU
Iridium ≥99.8% (metals basis), wire, Diameter 0.5 mm (0.02 in)
Iridium
Formula:
Ir MW: 192.22 g/mol Boiling Pt: 5300 °C (1013 hPa) Melting Pt: 2450 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011062 CAS Number: 7439-88-5 EINECS: 231-095-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...