Iridium ≥99.8% (metals basis), wire, Diameter 1.0 mm (0.04 in)
11429-BQ
11429-BO
011429-BQ
011429-BO
AA11429-BOEA
532.99
USD
AA11429-BO
CAAA11429-BQ
AA11429-BQ
CAAA11429-BO
Iridium ≥99.8% (metals basis), wire, Diameter 1.0 mm (0.04 in)
Iridium
Formula:
Ir MW: 192.22 g/mol Boiling Pt: 5300 °C (1013 hPa) Melting Pt: 2450 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011062 CAS Number: 7439-88-5 EINECS: 231-095-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...