Lead ≥99.9998% (metals basis), rods, Puratronic®, Diameter 12.7 mm (0.5 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
011346-97
011346-98
011346-18
11346-18
11346-97
11346-98
AA11346-18EA
104.27
USD
CAAA11346-18
AA11346-18
AA11346-97
CAAA11346-98
AA11346-98
CAAA11346-97
Lead ≥99.9998% (metals basis), rods, Puratronic®, Diameter 12.7 mm (0.5 in)
Lead
Formula:
Pb MW: 207.2 g/mol Boiling Pt: 1740 °C (1013 hPa) Melting Pt: 327 °C Density: 11.3437 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134050 CAS Number: 7439-92-1 EINECS: 231-100-4 Merck Index: 12,05410 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...