Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Thallium(I) acetate, Puratronic®, 99.9985% (metals basis)

Supplier: Alfa Aesar
Danger

11102-14 11102-22
MDL: MFCD00013045
Beilstein Registry No.: 3693914

Purity: 99.9985%
CAS No.: 563-68-8
Molecular Formula: Tl(OOCCH3)
Formula Weight: 263.42
Physical Form: Powder
Sensitivity: Hygroscopic
Beilstein Reference: 3693914
Merck Reference: 14,9257
UN No.: 1707
MDL No.: MFCD00013045


Formula: C₂H₃O₂Tl
MW: 263.43 g/mol
Boiling Pt: 1457 °C (1013 hPa)
Melting Pt: 110 °C
Density: 3.68 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013045
CAS Number: 563-68-8
EINECS: 209-257-5
UN: 1707
ADR: 6.1,II
Merck Index: 12,09392


Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Packaging Supplier No. VWR Catalog Number Unit Price Quantity
100 g bottle 11102-22 AA11102-22 Each (100g) Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Sensitivity Hygroscopic
Melting Point 131°C
Formula Weight 263.42
CAS No. 563-68-8
Purity 99.9985%
Physical Form Powder
Odor/Scent Odorless
Molecular Formula Tl(OOCCH3)
MDL No. MFCD00013045
UN No. 1707
Merck Reference 14,9257
Beilstein Reference 3693914
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty