Iron(III) sulfate ≥99.998% (metals basis), Puratronic®

Warning

Synonyms: Diiron trissulphate Ferric sulphate

10717-06 10717-06 10717-14
AA10717-06EA 130.74 USD
CAAA10717-14 CAAA10717-06 AA10717-06
Iron(III) sulfate ≥99.998% (metals basis), Puratronic®
Iron (III) sulphate
Formula: Fe₂O₁₂S₃
MW: 399.88 g/mol
Melting Pt: 480 °C (dec.)
Density: 3.097
Storage Temperature: Ambient
MDL Number: MFCD00011007
CAS Number: 10028-22-5
EINECS: 233-072-9
Merck Index: 12,04079

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR