Cobalt(II) fluoride tetrahydrate ≥99.99% (metals basis), Puratronic®
|
Danger
|
Synonyms:
Cobalt difluoride tetrahydrate
10693-06
10693-14
10693-06
010693-14
AA10693-14EA
235.87
USD
AA10693-14
CAAA10693-14
AA10693-06
CAAA10693-06
Cobalt(II) fluoride tetrahydrate ≥99.99% (metals basis), Puratronic®
Cobalt (II) fluoride tetrahydrate
Formula:
CoF₂·4H₂O MW: 168.99 g/mol Melting Pt: 200 °C (dec.) Density: 2.19 Storage Temperature: Ambient |
MDL Number:
MFCD00149656 CAS Number: 13817-37-3 EINECS: 233-061-9 UN: 2923 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...