Rhodium (III) acetylacetonate (≥25.2% Rh) ≥99.99% (metals basis), Premion®

Warning

Synonyms: Rhodium (III) 2,4-pentanedionate Rhodium (III) tris(2,4-pentanedionato-O,O') Tris(2,4-pentanedionato-O,O')rhodium (III)

10561-02 010561-06 010561-02 010561-03 10561-03 10561-06
AA10561-03EA 642.99 USD
AA10561-03 AA10561-02 CAAA10561-03 CAAA10561-02 AA10561-06 CAAA10561-06
Rhodium (III) acetylacetonate (≥25.2% Rh) ≥99.99% (metals basis), Premion®
Rhodium (III) acetylacetonate
Formula: C₁₅H₂₁O₆Rh
MW: 400.23 g/mol
Melting Pt: 263…264 °C
Storage Temperature: Ambient
MDL Number: MFCD00083144
CAS Number: 21679-46-9
EINECS: 238-192-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR