Hafnium ≥99.7% (trace metals basis excluding Zr), crystal bar milled chips (Zr ∼ 3%)
Supplier: Thermo Scientific Chemicals
10204-14
10204-22
010204-22
AA10204-22EA
664.99
USD
CAAA10204-22
CAAA10204-14
AA10204-22
Hafnium ≥99.7% (trace metals basis excluding Zr), crystal bar milled chips (Zr ∼ 3%)
Hafnium
Formula:
Hf MW: 178.49 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011032 CAS Number: 7440-58-6 EINECS: 231-166-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...