Calcium ≥99% (metals basis), shot ≤ 1 cm (0.4 in)

Danger

10126-22
AA10126-22EA 59.5 USD
AA10126-22
Calcium ≥99% (metals basis), shot ≤ 1 cm (0.4 in)
Calcium
Formula: Ca
MW: 40.08 g/mol
Boiling Pt: 1484 °C (1013 hPa)
Melting Pt: 850 °C
Density: 1.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085314
CAS Number: 7440-70-2
EINECS: 213-179-5
UN: 1401
ADR: 4.3,II
Merck Index: 13,01644

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR