Platinum rhodium alloy (70:30 w%), wire for ISA type B standard grade thermocouple, Diameter 0.25 mm (0.01 in)
10063-BY 10063-BW 10063-BW
AA10063-BWEA 372.95 USD
AA10063-BW CAAA10063-BW AA10063-BY
Platinum rhodium alloy (70:30 w%), wire for ISA type B standard grade thermocouple, Diameter 0.25 mm (0.01 in)
Platinum Rhodium alloy (70:30 w%)