Would you like to log in again?
Hoshizaki B-500PF Ice Bin
Supplier: NORDON/HOSHIZAKI LLC.
Hoshizaki
B-500PF
89166-546EA 2929.2 USD InStock
89166-546
Hoshizaki B-500PF Ice Bin
Ice Makers
Ice Bins are made of a vinyl clad exterior for easy cleaning and a long lastingattractive appearance
It's designed with a sturdy construction for side by side or stacked installation, and non-corrosive bin liner provides safe sanitary storage of ice cubes or flaked ice. Foamed-in-place polyurethane insulation in all bin walls, and bottom, provides dependable ice storage. The new baffle design increases ice accessibility by 30%. HoshiGuard Antimicrobal Agents add extra protection to the storage bin and ice scoop.
Certifications: Ice maker is AHRI and NSF certified.
Ordering information: Ice makers is supplied with manufacturer's two-year warranty on parts and labor.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty