Would you like to log in again?
Hoshizaki F-300BAF Ice Maker with Bin, Nordon Inc.
Supplier: NORDON/HOSHIZAKI LLC.
Hoshizaki
F-300BAF
89166-534EA 11962.52 USD InStock
89166-534
Hoshizaki F-300BAF Ice Maker with Bin, Nordon Inc.
Ice Makers
The Flaker/Cubelet Series features an attractive stainless steel finish
It contains an internal auger which is design to produce the highest quality of ice with fewer repairs and down time. The freezing cycle is made from cast stainless steel. The Flaker/Cubelet models also features a CleanCycle 12 design - 15 min flush every 12 hours. The self contained flakers flush automatically when the bin control is activated. These units are also the industries only greaseless bearing design to improve the life of the machine. This unit can produce up to 303lbs of ice per 24 hours.
Certifications: Ice maker is NSF certified and UL listed. Filters are AHRI and NSF certified.
Ordering information: Ice maker is supplied with manufacturer's three-year warranty on parts and labor and five-year warranty on compressor and air cooled condenser coil. 5 Year warranty on heads offered by Everpure.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty