Hoshizaki AM-50BAE Ice Maker with Bin, Nordon Inc.
Supplier: Hoshizaki
AM-50BAE
89166-514EA 5855.26 USD
89166-514
Hoshizaki AM-50BAE Ice Maker with Bin, Nordon Inc.
Ice Makers
The AM-50 undercounter design is small enough to fit conveniently into most cabinetry without additional ventilation.
  • Swing style reversible door and the UL outdoor approval expand the possibilities where the unit can be installed
  • Produces up to 55 lbs. of ice per 24 hour period with up to 27 lbs. of built in storage
  • Has fewer moving parts for a longer life
The power switch is easily accesible without having to remove the panel. You can easily disassembly the water circuit for easy cleaning.
115 V, 60 Hz, 1-Phase
Certifications: Ice maker is NSF certified and UL listed. Filters are AHRI and NSF certified.
Ordering information: Ice maker is supplied with two-year manufacturer's warranty on parts and labor. Filters are supplied with five-year manufacturer's warranty on heads offered by Everpure.