VWR Life Science Protein Molecular Weight Marker, Low Range
K880-200UL
97063-690EA 193.61 USD
97063-690
VWR Life Science Protein Molecular Weight Marker, Low Range
  • Biotechnology Grade
  • 6 bands ranging from 3.5 to 31.0 kDa
  • Excellent for gels separating low molecular weight proteins and polypeptides