Nonidet® P 40 Substitute (NP-40), Proteomics Grade

Warning

M158-500ML M158-50ML M158-100ML
97064-918EA 45.01 USD
97064-918 97064-920 97064-922
Nonidet® P 40 Substitute (NP-40), Proteomics Grade
Nonidet® P 40 Substitute (NP-40)

Non ionic detergent efficiently separates hydrophilic proteins from membrane spanning, hydrophobic proteins without altering biological activity.


  • High purity and protease-free
  • Suitable for most biological applications


Non-ionic detergents have uncharged hydrophilic groups and are gentle, non-denaturing detergents ideal for studying the conformation and function of proteins, because they keep secondary and tertiary structure intact.

Storage Temperature: Ambient
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR