Bromocresol green ACS

Supplier: VWR

0816-25G 0816-50G
97062-402EA 103.57 USD InStock
97062-402 97062-404
Bromocresol green ACS
Bromocresol green
Formula: C₂₁H₁₄Br₄O₅S
MW: 698.02 g/mol
Melting Pt: 217…218 °C
Storage Temperature: Ambient
MDL Number: MFCD00005874
CAS Number: 76-60-8
EINECS: 200-972-8
Merck Index: 13,01370
https://us.vwr.com/stibo/web/std.lang.all/13/36/2301336.jpg

Order Now

Specification Test Results

Clarity of Solution Pass
Visual Transition Interval (pH 3.8-5.4) Pass

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR