Tetramethylammonium chloride, high purity

Danger

0104-500G
97064-514CS 1744.3 USD
97064-514
Tetramethylammonium chloride, high purity
Tetramethylammonium chloride

TMA effectively abolishes the differential melting temperatures of AT and GC base pairs making it a useful reagent for nucleic acid hybridization procedures.

Formula: (CH₃)₄NCl
MW: 109.60 g/mol
Melting Pt: 368 °C…370 °C
MDL Number: MFCD00011628
CAS Number: 75-57-0
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Loss on Drying = 1 %
Purity = 99 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR