VWR® SSPE Buffer
Supplier: Avantor
0810-4L
97062-394EA 137.2 USD
97062-394
VWR® SSPE Buffer
SSPE Buffer is a hybridization buffer for Northern and Southern blots
VWR offers SSPE in various convenient forms: a 20X liquid concentrate, a tablet that prepares 100 mL of a 1X solution, and a Ready-Pack™ that prepares 1 L of 20X concentrate.