Deoxyribonucleotides are the foundation of DNA and it takes thousands of these nucleotides to make up DNA.
- Building blocks for the amplification of DNA
- High purity grade
DNA consists of a sugar molecule (deoxyribose), a nitrogenous base (purine in adenine and guanine, pyrimidine in thymine or cytosine) and a phosphate molecule. VWR’s deoxyribonucleotides are ideal for the amplification of DNA.
Ordering information: Kit includes:
dATP, 40 μmol of 100 mM Solution, pH 7.0
dTTP, 40 μmol of 100 mM Solution, pH 7.0
dCTP, 40 μmol of 100 mM Solution, pH 7.0
dGTP, 40 μmol of 100 mM Solution, pH 7.0