VWR®, dCTP
Supplier: Avantor
E536-40UM
97062-864EA 98.73 USD
97062-864
VWR®, dCTP
  • Building blocks for the amplification of DNA
  • High purity grade
Deoxyribonucleotides are the foundation of DNA and it takes thousands of these nucleotides to make up DNA. DNA consists of a sugar molecule (deoxyribose), a nitrogenous base (purine in adenine and guanine, pyrimidine in thymine or cytosine) and a phosphate molecule. VWR’s deoxyribonucleotides are ideal for the amplification of DNA.